Tris(8-hydroxyquinolinato)aluminum, 99%
Tris(8-hydroxyquinolinato)aluminum, 99%
Tris(8-hydroxyquinolinato)aluminum, 99%
Tris(8-hydroxyquinolinato)aluminum, 99%
Thermo Scientific Chemicals

Tris(8-hydroxyquinolinato)aluminum, 99%

CAS: 2085-33-8
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H55656.14
also known as H55656-14
Price (TWD)
-
Request A Quote
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS2085-33-8
IUPAC Name8-{[bis(quinolin-8-yloxy)alumanyl]oxy}quinoline
Molecular FormulaC27H18AlN3O3
InChI KeyTVIVIEFSHFOWTE-UHFFFAOYSA-K
SMILESO([Al](OC1=C2N=CC=CC2=CC=C1)OC1=C2N=CC=CC2=CC=C1)C1=C2N=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)98.5% min
Key green-light emitting and electron transport material for OLEDs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Key green-light emitting and electron transport material for OLEDs.

Solubility
Solubility in chloroform

Notes
Reacts with strong oxidizing agents. Store at room temperature.
RUO – Research Use Only

General References:

  1. Bo Chao Lin.; Cheu P. Cheng.; Zhi-Qiang You.; Chao-Ping Hsu. Charge Transport Properties of Tris(8-hydroxyquinolinato)aluminum(III): Why It Is an Electron Transporter.J. Am. Chem. Soc. 2005, 127 (1), 66-67.
  2. Gary P. Kushto.; Yasuhiro Iizumi.; Junji Kido.; Zakya H. Kafafi. A Matrix-Isolation Spectroscopic and Theoretical Investigation of Tris(8-hydroxyquinolinato)aluminum(III) and Tris(4-methyl-8-hydroxyquinolinato)aluminum(III).J. Phys. Chem. 2000, 104 (16),3670-3680.