1,10-Phenanthroline-5,6-dione, 98%
1,10-Phenanthroline-5,6-dione, 98%
1,10-Phenanthroline-5,6-dione, 98%
Thermo Scientific Chemicals

1,10-Phenanthroline-5,6-dione, 98%

CAS: 27318-90-7 | C12H6N2O2 | 210.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H55747.MD
also known as H55747-MD
Price (USD)
61.65
Special offer
Online exclusive
Ends: 30-Jun-2026
72.60
Save 10.95 (15%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
61.65
Special offer
Online exclusive
Ends: 30-Jun-2026
72.60
Save 10.95 (15%)
Each
Chemical Identifiers
CAS27318-90-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to yellow
FormPowder or crystals or crystalline powder
Assay from Suppliers CofA≥97.5%
1,10-Phenanthroline-5,6-dione is a Bifunctional quinone oxidant which, when used in conjunction with Zn2+ catalysts, is used to affect the aerobic oxidation of secondary amines to a variety of value added motifs, including indoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,10-Phenanthroline-5,6-dione is a Bifunctional quinone oxidant which, when used in conjunction with Zn2+ catalysts, is used to affect the aerobic oxidation of secondary amines to a variety of value added motifs, including indoles.

Solubility
Insoluble in water.

Notes
Store in a cool and dark place. Store away from incompatible materials such as oxidizing agents.
RUO – Research Use Only

General References:

  1. Charles A. Goss.; Hector D. Abruna. Spectral, electrochemical and electrocatalytic properties of 1,10-phenanthroline-5,6-dione complexes of transition metals. Inorg. Chem. 1985, 24 (25), 4263-4267.
  2. Q. Wu.; M. Maskus.; F. Pariente.; F. Tobalina.; V. M. Fernández.; E. Lorenzo.; H. D. Abruña. Electrocatalytic Oxidation of NADH at Glassy Carbon Electrodes Modified with Transition Metal Complexes Containing 1,10-Phenanthroline-5,6-dione Ligands. Anal. Chem. 1996, 68 (20),3688-3696.