2-Chloro-4-hydroxybenzaldehyde, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Chloro-4-hydroxybenzaldehyde, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in organic solvents.
Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only
General References:
- H Wynberg. The Reimer-Tiemann Reaction. Chemical Reviews. 200660 (2), 169-184.
- A Núñez-Montenegro.; R Carballo.; EM Vázquez-López. Synthesis and characterization of thiosemicarbazone derivatives of 2-chloro-4-hydroxy-benzaldehyde and their rhenium (I) complexes. Polyhedron. 200928 (18), 3915-3922.