3-Chloropyridine-4-carboxaldehyde, 98%
3-Chloropyridine-4-carboxaldehyde, 98%
3-Chloropyridine-4-carboxaldehyde, 98%
3-Chloropyridine-4-carboxaldehyde, 98%
Thermo Scientific Chemicals

3-Chloropyridine-4-carboxaldehyde, 98%

CAS: 72990-37-5 | C6H4ClNO | 141.554 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H55852.03
also known as H55852-03
Price (EUR)
43,90
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
43,90
Each
Add to cart
Chemical Identifiers
CAS72990-37-5
IUPAC Name3-chloropyridine-4-carbaldehyde
Molecular FormulaC6H4ClNO
InChI KeyZVGDKOQPJCOCLI-UHFFFAOYSA-N
SMILESClC1=C(C=O)C=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only