4-tert-Butylphenyl glycidyl ether, 95%
4-tert-Butylphenyl glycidyl ether, 95%
4-tert-Butylphenyl glycidyl ether, 95%
4-tert-Butylphenyl glycidyl ether, 95%
Thermo Scientific Chemicals

4-tert-Butylphenyl glycidyl ether, 95%

CAS: 3101-60-8 | C13H18O2 | 206.285 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H55945.06
also known as H55945-06
Price (TWD)
-
Request A Quote
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS3101-60-8
IUPAC Name2-[(4-tert-butylphenoxy)methyl]oxirane
Molecular FormulaC13H18O2
InChI KeyHHRACYLRBOUBKM-UHFFFAOYNA-N
SMILESCC(C)(C)C1=CC=C(OCC2CO2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>94.0%
4-tert-Butylphenyl glycidyl ether is used in paint industry and serves as organic coating in Paint, boat Paint, building coating material. It is also used as an epoxy reactive diluent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-Butylphenyl glycidyl ether is used in paint industry and serves as organic coating in Paint, boat Paint, building coating material. It is also used as an epoxy reactive diluent.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Q Fu.; W Lin.; J Huang . A New Strategy for Preparation of Graft Copolymers via Graft onto by Atom Transfer Nitroxide Radical Coupling Chemistry: Preparation of Poly(4-glycidyloxy-2,2,6,6-tetramethylpiperidine-1-oxyl-co-ethylene oxide)-graft-polystyrene and Poly(tert-butyl acrylate). Macromolecules.200841 (7), 2381-2387.
  2. K Taguchi.; S Hirose.; Y Abe. Photo-curing composite paint containing urushi (Oriental lacquer), and wrinkled coating caused by phase separation. Progress in organic coatings.200758 (4), 290-295.