Dihidrato de tetrakis(4-fluorofenil)borato de sodio, 98 %, Thermo Scientific Chemicals
Dihidrato de tetrakis(4-fluorofenil)borato de sodio, 98 %, Thermo Scientific Chemicals
Dihidrato de tetrakis(4-fluorofenil)borato de sodio, 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dihidrato de tetrakis(4-fluorofenil)borato de sodio, 98 %, Thermo Scientific Chemicals

CAS: 207683-22-5 | C24H20BF4NaO2 | 450.215 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
250 mg
5 g
Número de catálogo H55952.MD
Precio (USD)
-
Cantidad:
250 mg
Pedido a granel o personalizado
Identificadores químicos
CAS207683-22-5
EspecificacionesSpecification SheetHoja de especificaciones
FormCrystals or powder or crystalline powder
Assay from Supplier's CofA≥97.5% (HPLC)
Appearance (Color)Colorless, translucent or white
Sodium tetrakis(4-fluorophenyl)borate dihydrate, is used as a reagent for the two-phase titration of nonionic surfactants in the presence of anionic surfactants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Aplicaciones
El dihidrato de tetrakis(4-fluorofenil)borato de sodio se utiliza como reactivo en la valoración bifásica de los tensioactivos no iónicos en presencia de tensioactivos aniónicos.

Notas
Higroscópico. Incompatible con agentes oxidantes fuertes.
RUO – Research Use Only

General References:

  1. Funada, T.; Hirose, T.; Tamai, N.; Yao, H. Organic nanoparticles of malachite green with enhanced far-red emission: size-dependence of particle rigidity. Phys. Chem. Chem. Phys. 2015, 17 (16), 11006-11013.
  2. Lisak, G.; Arnebrant, T.; Ruzgas, T.; Bobacka, J. Textile-based sampling for potentiometric determination of ions. Anal. Chim. Acta 2015, 877, 71-79.