1-(1-Naphthyl)-2-(trimethylsilyl)acetylene, 97%
1-(1-Naphthyl)-2-(trimethylsilyl)acetylene, 97%
1-(1-Naphthyl)-2-(trimethylsilyl)acetylene, 97%
Thermo Scientific Chemicals

1-(1-Naphthyl)-2-(trimethylsilyl)acetylene, 97%

CAS: 104784-51-2 | C15H16Si | 224.378 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H55953.14
also known as H55953-14
Price (EUR)
551,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
551,00
Each
Specifications
CAS104784-51-2
Chemical Name or Material1-(1-Naphthyl)-2-(trimethylsilyl)acetylene
Density0.968 g/mL
Flash Point>110°C (230°F)
Formula Weight224.38
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only