Thermo Scientific Chemicals

n-Undecanoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: H56380.ME
500 mg, Each
Thermo Scientific Chemicals

n-Undecanoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: H56380.ME
500 mg, Each
Quantity
Catalog number: H56380.ME
also known as H56380-ME
Price (USD)
Price: 110.00
Online price: 93.65
Your price:
Quantity
-

Chemical Identifiers

CAS
17746-05-3
IUPAC Name
undecanoyl chloride
Molecular Formula
C11H21ClO
InChI Key
JUKPJGZUFHCZQI-UHFFFAOYSA-N
SMILES
CCCCCCCCCCC(Cl)=O
Assay (GC)
98%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text