Thermo Scientific Chemicals

Sodium phenylpyruvate, 98%, Thermo Scientific Chemicals

Catalog number: H56767.06
5 g, Each
Thermo Scientific Chemicals

Sodium phenylpyruvate, 98%, Thermo Scientific Chemicals

Catalog number: H56767.06
5 g, Each
Quantity
Catalog number: H56767.06
also known as H56767-06
Price (USD)
Price: 51.70
Online price: 43.65
Your price:
Quantity
-

Chemical Identifiers

CAS
114-76-1
IUPAC Name
sodium 2-oxo-3-phenylpropanoate
Molecular Formula
C9H7NaO3
InChI Key
MQGYVGKMCRDEAF-UHFFFAOYSA-M
SMILES
[Na+].[O-]C(=O)C(=O)CC1=CC=CC=C1
Assay (unspecified)
>97.5%

Description

Phenylpyruvic acid, sodium salt is a substrate for phenylpyruvate decarboxylase and phenylpyruvate tautomerase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenylpyruvic acid, sodium salt is a substrate for phenylpyruvate decarboxylase and phenylpyruvate tautomerase.

Solubility
Soluble in water

Notes
Store at 2-8°C. Avoin oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text