3-Aminooxetane-3-carboxylic acid, 95%
3-Aminooxetane-3-carboxylic acid, 95%
3-Aminooxetane-3-carboxylic acid, 95%
3-Aminooxetane-3-carboxylic acid, 95%
Thermo Scientific Chemicals

3-Aminooxetane-3-carboxylic acid, 95%

CAS: 138650-24-5 | C4H7NO3 | 117.104 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
500 mg
Catalog number H57291.ME
also known as H57291-ME
Price (EUR)
240,00
Each
Add to cart
Quantity:
500 mg
Request bulk or custom format
Price (EUR)
240,00
Each
Add to cart
Specifications
CAS138650-24-5
Chemical Name or Material3-Aminooxetane-3-carboxylic acid
ColorWhite
Formula Weight117.1
Health Hazard 1H302-H319
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only