3-Oxetanyl p-toluenesulfonate, 96%, Thermo Scientific Chemicals
3-Oxetanyl p-toluenesulfonate, 96%, Thermo Scientific Chemicals
3-Oxetanyl p-toluenesulfonate, 96%, Thermo Scientific Chemicals
3-Oxetanyl p-toluenesulfonate, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Oxetanyl p-toluenesulfonate, 96%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H57569.06
also known as H57569-06
5 g
Catalog number H57569.06
also known as H57569-06
Price (TWD)
-
Request A Quote
Quantity:
5 g
Request bulk or custom format
Specifications
CAS26272-83-3
Chemical Name or Material3-Oxetanyl p-toluenesulfonate
Density1.34
Flash Point179°C (354°F)
Formula Weight228.27
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only