Oxetane-3-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Oxetane-3-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Slightly soluble in water.
Notes
Store in a cool place. Moisture sensitive. . Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
- de Vicente, J.; Tivitmahaisoon, P.; Berry, P.; Bolin, D. R.; Carvajal, D.; He, W.; Huang, K. S.; Janson, C.; Liang, L.; Lukacs, C.; Petersen, A.; Qian, H.; Yi, L.; Zhuang, Y.; Hermann, J. C. Fragment-Based Drug Design of Novel Pyranopyridones as Cell Active and Orally Bioavailable Tankyrase Inhibitors. ACS Med. Chem. Lett. 2015, 6 (9), 1019-1024.
- Roydhouse, M. D.; Motherwell, W. B.; Constantinou, A.; Gavriilidis, A.; Wheeler, R.; Down, K.; Campbell, I. Ozonolysis of some complex organic substrates in flow. RSC Adv. 2013, 3 (15), 5076-5082.