6-Oxa-1-azaspiro[3.3]heptane hemioxalate, 95%, Thermo Scientific Chemicals
6-Oxa-1-azaspiro[3.3]heptane hemioxalate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Oxa-1-azaspiro[3.3]heptane hemioxalate, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
250 mg
Catalog number H57658.ME
also known as H57658-ME
Price (USD)/ Each
696.65
Online exclusive
819.00 
Save 122.35 (15%)
-
Add to cart
Quantity:
500 mg
Request bulk or custom format
Price (USD)/ Each
696.65
Online exclusive
819.00 
Save 122.35 (15%)
Add to cart
6-Oxa-1-azaspiro[3.3]heptane hemioxalate, 95%, Thermo Scientific Chemicals
Catalog numberH57658.ME
Price (USD)/ Each
696.65
Online exclusive
819.00 
Save 122.35 (15%)
-
Add to cart
Specifications
Chemical Name or Material6-Oxa-1-azaspiro[3.3]heptane hemioxalate
CAS1380571-72-1
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Notes
Incompatible with strong oxidizing agents, strong acids and bases.
RUO – Research Use Only
Burkhard, J. A.; Guérot, C.; Knust, H.; Rogers-Evans, M.; Carreira, E. M. Synthesis and Structural Analysis of a New Class of Azaspiro[3.3]heptanes as Building Blocks for Medicinal Chemistry. Org. Lett. 2010, 12 (9), 1944-1947.