N-Boc-D-beta-proline, 95%, Thermo Scientific Chemicals
N-Boc-D-beta-proline, 95%, Thermo Scientific Chemicals
N-Boc-D-beta-proline, 95%, Thermo Scientific Chemicals
N-Boc-D-beta-proline, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-D-beta-proline, 95%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H57825.03
also known as H57825-03
1 g
Catalog number H57825.03
also known as H57825-03
Price (EUR)
160,00
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
160,00
Each
Add to cart
Specifications
CAS72925-16-7
Chemical Name or MaterialN-Boc-D-beta-proline
Formula Weight215.25
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Soluble in Dimethyl sulfoxide (DMSO).

Notes
Store in a cool ventilated place. Avoid strong oxidizing materials. Hygroscopic.
RUO – Research Use Only
David Alker.; Kevin J. Doyle.; Laurence M. Harwood.; Andrew McGregor. The direct synthesis of the cyclic sulphamidate of (S)-prolinol: SimultaneousN-protection and activation towards nucleophilic displacement of oxygen. Tetrahedron: Asymmetry. 1990, 1 (12),877-880.