2-n-Propyl-1-pentanol, 97%
2-n-Propyl-1-pentanol, 97%
2-n-Propyl-1-pentanol, 97%
Thermo Scientific Chemicals

2-n-Propyl-1-pentanol, 97%

CAS: 58175-57-8 | C8H18O | 130.231 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H57954.03
also known as H57954-03
Price (EUR)
45,80
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
45,80
Each
Chemical Identifiers
CAS58175-57-8
IUPAC Name2-propylpentan-1-ol
Molecular FormulaC8H18O
InChI KeyLASHFHLFDRTERB-UHFFFAOYSA-N
SMILESCCCC(CO)CCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Purity≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only