3-(Ethoxycarbonyl)propylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles
3-(Ethoxycarbonyl)propylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles
3-(Ethoxycarbonyl)propylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles
3-(Ethoxycarbonyl)propylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles
Thermo Scientific Chemicals

3-(Ethoxycarbonyl)propylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

CAS: 131379-39-0 | C6H11BrO2Zn | 260.436 g/mol
Have Questions?
Catalog NumberQuantity
H58023.AD
also known as H58023-AD
50 mL
Catalog number H58023.AD
also known as H58023-AD
Price (EUR)
174,00
Each
Add to cart
Quantity:
50 mL
Request bulk or custom format
Price (EUR)
174,00
Each
Add to cart
Chemical Identifiers
CAS131379-39-0
IUPAC Name4-ethoxy-4-oxobutan-1-ide; bromozincylium
Molecular FormulaC6H11BrO2Zn
InChI KeyXPARCVGSTPKNNR-UHFFFAOYSA-M
SMILES[Zn+]Br.CCOC(=O)CC[CH2-]
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid, packaged under Argon in resealable ChemSeal™ bottles.
Assay from Suppliers CofA0.4-0.6M in THF
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Palladium-catalyzed coupling with [4-(methoxycarbonyl)benzyllzinc bromide then selective hydrolysis of the tert-butyl ester trifluoroacetic acid, in design and synthesis of pyridone-containing inhibitors.

Solubility
RUO – Research Use Only
Ilane Marek.; Jean-F. Normant. Synthesis and Reactivity of sp3-Geminated Organodimetallics. Chem. Rev. 1996, 96 (8),3241-3268.