4-(Ethoxycarbonyl)butylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles
4-(Ethoxycarbonyl)butylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles
4-(Ethoxycarbonyl)butylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles
Thermo Scientific Chemicals

4-(Ethoxycarbonyl)butylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

CAS: 131379-16-3 | C9H9IO2Zn | 341.45 g/mol
Have Questions?
Catalog NumberQuantity
H58484.AD
also known as H58484-AD
50 mL
Catalog number H58484.AD
also known as H58484-AD
Price (EUR)
-
Quantity:
50 mL
Request bulk or custom format
Specifications
CAS131379-16-3
Chemical Name or Material4-(Ethoxycarbonyl)phenylzinc iodide
DOT InformationTransport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
Density1.023
Formula Weight341.45
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-(Ethoxycarbonyl)phenylzinc iodide is used as pharmaceutical intermediates.

Solubility
Reacts with water.

Notes
Store at 4°C. Air Sensitive. Store under dry inert gas. Store away from air. Inco
RUO – Research Use Only
Laurin Melzig; Albrecht Metzger; Paul Knochel. Pd- and Ni-Catalyzed Cross-Coupling Reactions of Functionalized Organozinc Reagents with Unsaturated Thioethers. Chemistry - A European Journal.2011, 17 2948-2956.