2-Bromobenzylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles
2-Bromobenzylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles
2-Bromobenzylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles
Thermo Scientific Chemicals

2-Bromobenzylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

CAS: 307496-27-1 | C7H6Br2Zn | 315.313 g/mol
Have Questions?
Catalog NumberQuantity
H58678.AD
also known as H58678-AD
50 mL
Catalog number H58678.AD
also known as H58678-AD
Price (EUR)
-
Quantity:
50 mL
Request bulk or custom format
Specifications
CAS307496-27-1
Chemical Name or Material2-Bromobenzylzinc bromide
DOT InformationTransport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
Density1.015
Formula Weight315.31
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only