Yoduro de 1-etil-3-metilimidazolio, 97 %, Thermo Scientific Chemicals
Yoduro de 1-etil-3-metilimidazolio, 97 %, Thermo Scientific Chemicals
Yoduro de 1-etil-3-metilimidazolio, 97 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Yoduro de 1-etil-3-metilimidazolio, 97 %, Thermo Scientific Chemicals

CAS: 35935-34-3 | C6H11IN2 | 238.07 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
5 g
50 g
Número de catálogo H59404.18
también denominado H59404-18
Precio (EUR)
187,00
Each
Cantidad:
50 g
Pedido a granel o personalizado
Precio (EUR)
187,00
Each
Identificadores químicos
CAS35935-34-3
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)White to pale yellow
Proton NMR>96.0%
FormPowder
It is used as OLED materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Aplicaciones
Se utiliza como material OLED.

Solubilidad
Soluble en agua, diclorometano, etanol y acetonitrilo.

Notas
Sensible a la humedad. Mantener alejado del calor y la humedad. Incompatible con agentes oxidantes fuertes.
RUO – Research Use Only

General References:

  1. Annegret Stark.; Bonnie L. MacLean.; Robert D. Singer. 1-Ethyl-3-methylimidazolium halogenoaluminate ionic liquids as solvents for Friedel-Crafts acylation reactions of ferrocene. J. Chem. Soc., Dalton Trans. 1999, (1),63-66.
  2. Laine Green.; Ivan Hemeon.; Robert D Singer. 1-Ethyl-3-methylimidazolium halogenoaluminate ionic liquids as reaction media for the acylative cleavage of ethers.Tetrahedron Lett. 2000, 41 (9), 1343-1346.