1,3,5-Tri-O-benzoyl-alpha-D-ribofuranose, 97%, Thermo Scientific Chemicals
1,3,5-Tri-O-benzoyl-alpha-D-ribofuranose, 97%, Thermo Scientific Chemicals
1,3,5-Tri-O-benzoyl-alpha-D-ribofuranose, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,3,5-Tri-O-benzoyl-alpha-D-ribofuranose, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H59464.06
also known as H59464-06
5 g
Catalog number H59464.06
also known as H59464-06
Price (EUR)
183,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
183,00
Each
Chemical Identifiers
CAS22224-41-5
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
It is employed as a starting material for nucleosides and is an intermediate fir cloforabine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a starting material for nucleosides and is an intermediate fir cloforabine.

Solubility
Insoluble in water, easily soluble in chloroform.

Notes
Store at room temperature. Keep away from oxidizing agents and heats.
RUO – Research Use Only
Paul R. Brodfuehrer.; Chester SapinoJr.; Henry G. Howell. A stereocontrolled synthesis of 1,3,5-tri-O-benzoyl-.alpha.-D-ribofuranose. J. Org. Chem. 1985, 50 (14),2597-2598.