6-Bromoquinoline-5-carbonitrile, 95%
6-Bromoquinoline-5-carbonitrile, 95%
6-Bromoquinoline-5-carbonitrile, 95%
Thermo Scientific Chemicals

6-Bromoquinoline-5-carbonitrile, 95%

CAS: 1188365-70-9 | C10H5BrN2 | 233.068 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H59484.03
also known as H59484-03
Price (EUR)
171,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
171,00
Each
Specifications
Beilstein20114547
CAS1188365-70-9
Chemical Name or Material6-Bromoquinoline-5-carbonitrile
DOT InformationTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Formula Weight233.07
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only