Thermo Scientific Chemicals

L-Glutamic acid diethyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: H59551.14
25 g, Each
Thermo Scientific Chemicals

L-Glutamic acid diethyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: H59551.14
25 g, Each
Quantity
Catalog number: H59551.14
also known as H59551-14
Price (USD)
Price: 121.00
Online price: 104.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1118-89-4
IUPAC Name
hydrogen 1,5-diethyl (2S)-2-aminopentanedioate chloride
Molecular Formula
C9H18ClNO4
InChI Key
WSEQLMQNPBNMSL-FJXQXJEOSA-N
SMILES
[H+].[Cl-].CCOC(=O)CC[C@H](N)C(=O)OCC
Proton NMR
>97.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text