Tin(II) methanesulfonate, 50% w/w aq. soln.
Tin(II) methanesulfonate, 50% w/w aq. soln.
Tin(II) methanesulfonate, 50% w/w aq. soln.
Thermo Scientific Chemicals

Tin(II) methanesulfonate, 50% w/w aq. soln.

CAS: 53408-94-9 | C2H6O6S2Sn | 308.89 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
Catalog number H59773.AK
also known as H59773-AK
Price (EUR)
68,40
Each
Quantity:
250 mL
Request bulk or custom format
Price (EUR)
68,40
Each
Chemical Identifiers
CAS53408-94-9
IUPAC Nameλ²-tin(2+) dimethanesulfonate
Molecular FormulaC2H6O6S2Sn
InChI KeyJALQQBGHJJURDQ-UHFFFAOYSA-L
SMILES[Sn++].CS([O-])(=O)=O.CS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration)≥50.0%
Appearance (Color)Clear colorless to pale yellow
FormLiquid
It is used in electroplating industry. Tin methanesulfonate is an aqueous solution used in the metal surface treatment. Mainly as a replacement for Tin sulfate where high speed deposits are needed (reel-to-reel).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in electroplating industry. Tin methanesulfonate is an aqueous solution used in the metal surface treatment. Mainly as a replacement for Tin sulfate where high speed deposits are needed (reel-to-reel).

Solubility
Not miscible or difficult to mix in water.

Notes
Air sensitive. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Kok Kee Yang, et al. Diffusion Coefficient of Tin(II) Methanesulfonate in Ionic Liquid and Methane Sulfonic Acid (MSA) Solvent.Metall. Mater. Trans. B.,2011,42(6), 1274-1279.
  2. Algirdas Vaškelis, et al. Silver nanostructured catalyst for modification of dielectrics surface.Electrochim. Acta.,2005,50(23), 4586-4591.