6-Amino-7-bromo-9-methyl-7-deazapurine, 97%, Thermo Scientific Chemicals
6-Amino-7-bromo-9-methyl-7-deazapurine, 97%, Thermo Scientific Chemicals
6-Amino-7-bromo-9-methyl-7-deazapurine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Amino-7-bromo-9-methyl-7-deazapurine, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H59897.03
also known as H59897-03
1 g
Catalog number H59897.03
also known as H59897-03
Price (EUR)
338,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
338,00
Each
Specifications
Beilstein21981671
CAS1337532-51-0
Chemical Name or Material6-Amino-7-bromo-9-methyl-7-deazapurine
Formula Weight227.06
Health Hazard 1H315-H319-H335
View more
It is used in the preparation of indoline derivatives as PERK inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the preparation of indoline derivatives as PERK inhibitors.

Solubility
Soluble in dichloromethane.

Notes
Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only
Stefan A. Laufer.; David M. Domeyer.; Thomas R. F. Scior.; Wolfgang Albrech.; Dominik R. J. Hauset. Synthesis and Biological Testing of Purine Derivatives as Potential ATP-Competitive Kinase Inhibitors. J. Med. Chem. 2005, 48 (3),710-722.