4-Hydroxybenzenesulfonic acid, 65%
4-Hydroxybenzenesulfonic acid, 65%
4-Hydroxybenzenesulfonic acid, 65%
Thermo Scientific Chemicals

4-Hydroxybenzenesulfonic acid, 65%

CAS: 98-67-9 | C6H6O4S | 174.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Catalog number H59917.AK
also known as H59917-AK
Price (EUR)
68,20
Each
Quantity:
250 mL
Request bulk or custom format
Price (EUR)
68,20
Each
Chemical Identifiers
CAS98-67-9
IUPAC Name4-hydroxybenzene-1-sulfonic acid
Molecular FormulaC6H6O4S
InChI KeyFEPBITJSIHRMRT-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Concentration (weight/weight)65 wt. % in H2O
4-Hydroxybenzenesulfonic Acid is used as a redox-mediator in the laccase-catalyzed degradation of indigo, a textile dye. Also used in oilfield chemicals, metal working, and as a polymerization catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxybenzenesulfonic Acid is used as a redox-mediator in the laccase-catalyzed degradation of indigo, a textile dye. Also used in oilfield chemicals, metal working, and as a polymerization catalyst.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. R Campos; A Kandelbauer; K.H Robra; Artur Cavaco-Paulo; G.M Gübitz. Indigo degradation with purified laccases from Trametes hirsuta and Sclerotium rolfsii. Journal of Biotechnology,2001, 89 (2-3), 131-139.
  2. A M Lambeir; H B Dunford; M A Pickard. Kinetics of the oxidation of ascorbic acid, ferrocyanide and p-phenolsulfonic acid by chloroperoxidase compounds I and II. European Journal of Biochemistry. 1987, 163(1), 123-127.