1,3,5-Tri(4-carboxyphenyl)benzene, 97%
1,3,5-Tri(4-carboxyphenyl)benzene, 97%
1,3,5-Tri(4-carboxyphenyl)benzene, 97%
Thermo Scientific Chemicals

1,3,5-Tri(4-carboxyphenyl)benzene, 97%

CAS: 50446-44-1 | C27H18O6 | 438.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H60087.03
also known as H60087-03
Price (USD)
112.65
Online Exclusive
125.00
Save 12.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
112.65
Online Exclusive
125.00
Save 12.35 (10%)
Each
Chemical Identifiers
CAS50446-44-1
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Proton NMRConforms to structure
Appearance (Color)White to pale cream
Assay from Supplier's CofA≥96.0%
1,3,5-Tri(4-carboxyphenyl)benzene is used as a building block for metal organic frameworks, which is a 3D-microporous material and finds applications in gas adsorption and separation technologies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Tri(4-carboxyphenyl)benzene is used as a building block for metal organic frameworks, which is a 3D-microporous material and finds applications in gas adsorption and separation technologies.

Solubility
Soluble in tetrahydrofuran.

Notes
Incompatible with strong oxidizing agents, metals, amines, strong acids, acid chlorides, alcohols, peroxides, permanganates, strong reducing agents, soluble carbonates, phosphates, phosphorus halides and hydroxides.
RUO – Research Use Only

General References:

  1. Li, D. X.; Ren, Z. G.; Young, D. J.; Lang, J. P. Synthesis of Two Coordination Polymer Photocatalysts and Significant Enhancement of Their Catalytic Photodegradation Activity by Doping with Co2+ Ions. Eur. J. Inorg. Chem. 2015, 2015 (11), 1981-1988.
  2. Clough, A.; Zheng, S. T.; Zhao, X.; Lin, Q.; Feng, P.; Bu, X. New Lithium Ion Clusters for Construction of Porous MOFs. Cryst. Growth Des. 2014, 14 (3), 897-900.