Thermo Scientific Chemicals

Phenylazoformic acid 2-phenylhydrazide, Thermo Scientific Chemicals

Catalog number: H60336.14
25 g, Each
Thermo Scientific Chemicals

Phenylazoformic acid 2-phenylhydrazide, Thermo Scientific Chemicals

Catalog number: H60336.14
25 g, Each
Quantity
Catalog number: H60336.14
also known as H60336-14
Price (USD)
Price: 310.00
Online price: 268.65
Your price:
Quantity
-

Chemical Identifiers

CAS
538-62-5
IUPAC Name
3-(phenylamino)-1-(phenylimino)urea
Molecular Formula
C13H12N4O
InChI Key
ZFWAHZCOKGWUIT-UHFFFAOYSA-N
SMILES
O=C(NNC1=CC=CC=C1)N=NC1=CC=CC=C1
Appearance (Color)
Orange to red to brown
Form
Crystals or crystalline powder or powder
Melting Point
154-161?C
Residue on ignition
≤0.1%

Description

It is a reagent for mercury determination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a reagent for mercury determination.

Solubility
Soluble in acetone. Insoluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text