1H-1,2,4-Triazole-1-carboxamidine hydrochloride, 98%
1H-1,2,4-Triazole-1-carboxamidine hydrochloride, 98%
1H-1,2,4-Triazole-1-carboxamidine hydrochloride, 98%
1H-1,2,4-Triazole-1-carboxamidine hydrochloride, 98%
Thermo Scientific Chemicals

1H-1,2,4-Triazole-1-carboxamidine hydrochloride, 98%

CAS: 19503-26-5 | C3H6ClN5 | 147.566 g/mol
Have Questions?
Catalog NumberQuantity
H60475.06
also known as H60475-06
5 g
Catalog number H60475.06
also known as H60475-06
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS19503-26-5
IUPAC Name1H-1,2,4-triazole-1-carboximidamide hydrochloride
Molecular FormulaC3H6ClN5
InChI KeyJDDXNENZFOOLTP-UHFFFAOYSA-N
SMILESCl.NC(=N)N1C=NC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only