1,2-Benzisothiazol-3-one, 97%
1,2-Benzisothiazol-3-one, 97%
1,2-Benzisothiazol-3-one, 97%
Thermo Scientific Chemicals

1,2-Benzisothiazol-3-one, 97%

CAS: 2634-33-5 | C7H5NOS | 151.183 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H60492.06
also known as H60492-06
Price (EUR)
113,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
113,00
Each
Chemical Identifiers
CAS2634-33-5
IUPAC Name2,3-dihydro-1,2-benzothiazol-3-one
Molecular FormulaC7H5NOS
InChI KeyDMSMPAJRVJJAGA-UHFFFAOYSA-N
SMILESO=C1NSC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream or pale yellow
FormPowder or crystalline powder
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Melting Point154-163°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
It is an antimicrobial agent and a pharmaceutical intermediate.

Solubility
Soluble in dichloromethane, dimethyl sulfoxide, methanol.

Notes
Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Keith D. Combrink .; H. Belgin Gülgeze.; Nicholas A. Meanwell .; Bradley C. Pearce.; Pi Zulan .; Gregory S. Bisacchi.; Daniel G. M. Roberts.; Paul Stanley.; Steven M. Seiler. 1,2-Benzisothiazol-3-one 1,1-Dioxide Inhibitors of Human Mast Cell Tryptase. J. Med. Chem. 1998, 41 (24),4854-4860.
  2. Magid Abou-Gharbia.; John A. Moyer.; Usha Patel.; Michael Webb.; Guy Schiehser.; Terrance Andree.; J. Thomas Haskins. Synthesis and structure-activity relationship of substituted tetrahydro- and hexahydro-1,2-benzisothiazol-3-one 1,1-dioxides and thiadiazinones: potential anxiolytic agents. J. Med. Chem. 1989, 32 (5),1024-1033.