Bis[2-(di-tert-butylphosphino)ethyl]amine, 10% w/w soln. in THF
Bis[2-(di-tert-butylphosphino)ethyl]amine, 10% w/w soln. in THF
Bis[2-(di-tert-butylphosphino)ethyl]amine, 10% w/w soln. in THF
Bis[2-(di-tert-butylphosphino)ethyl]amine, 10% w/w soln. in THF
Thermo Scientific Chemicals

Bis[2-(di-tert-butylphosphino)ethyl]amine, 10% w/w soln. in THF

CAS: 944710-34-3 | C20H45NP2 | 361.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H60493.03
also known as H60493-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Specifications
CAS944710-34-3
Chemical Name or MaterialBis[2-(di-tert-butylphosphino)ethyl]amine
Concentration or Composition (by Analyte or Components)10% w/w soln. in THF
DOT InformationTransport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Density0.911
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
This compound is involved in the transition metal-catalyzed addition of amines to acrylic acid derivatives.

Solubility
Insoluble in water.

Notes
Air Sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only
Motoi Kawatsura.; John F. Hartwig. Transition Metal-Catalyzed Addition of Amines to Acrylic Acid Derivatives. A High-Throughput Method for Evaluating Hydroamination of Primary and Secondary Alkylamines.Organometallics. 2001, 20 (10),1960-1964.