2-Methyl-2-propen-1-ol, 98%
2-Methyl-2-propen-1-ol, 98%
2-Methyl-2-propen-1-ol, 98%
Thermo Scientific Chemicals

2-Methyl-2-propen-1-ol, 98%

CAS: 513-42-8 | C4H8O | 72.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number H60502.06
also known as H60502-06
Price (USD)
46.65
Special offer
Online exclusive
Ends: 30-Jun-2026
54.20
Save 7.55 (14%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
46.65
Special offer
Online exclusive
Ends: 30-Jun-2026
54.20
Save 7.55 (14%)
Each
Chemical Identifiers
CAS513-42-8
IUPAC Name2-methylprop-2-en-1-ol
Molecular FormulaC4H8O
InChI KeyBYDRTKVGBRTTIT-UHFFFAOYSA-N
SMILESCC(=C)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)97.5% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
 
RUO – Research Use Only