(R,R)-(-)-2-Amino-1-phenylpropyldiphenylphosphine, 97+%
(R,R)-(-)-2-Amino-1-phenylpropyldiphenylphosphine, 97+%
(R,R)-(-)-2-Amino-1-phenylpropyldiphenylphosphine, 97+%
Thermo Scientific Chemicals

(R,R)-(-)-2-Amino-1-phenylpropyldiphenylphosphine, 97+%

CAS: 799297-44-2 | C21H22NP | 319.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H60639.MD
Price (EUR)
127,00
Each
Quantity:
250 mg
Request bulk or custom format
Price (EUR)
127,00
Each
Specifications
CAS799297-44-2
Chemical Name or Material(R,R)-(-)-2-Amino-1-phenylpropyldiphenylphosphine
DOT InformationTransport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
Formula Weight319.38
Health Hazard 1H314-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only