規格
CAS1365531-82-3
Chemical Name or Material(S)-2,2'-Bis(di-p-tolylphosphino)-4,4',6,6'-tetramethoxybiphenyl
Formula Weight698.77
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
檢視更多