(S)-1-(2-Chlorophenyl)ethanol, 98%
(S)-1-(2-Chlorophenyl)ethanol, 98%
(S)-1-(2-Chlorophenyl)ethanol, 98%
Thermo Scientific Chemicals

(S)-1-(2-Chlorophenyl)ethanol, 98%

CAS: 131864-71-6 | C8H9ClO | 156.609 g/mol
Have Questions?
Catalog NumberQuantity
H60935.03
also known as H60935-03
1 g
Catalog number H60935.03
also known as H60935-03
Price (TWD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS131864-71-6
IUPAC Name1-(2-chlorophenyl)ethan-1-ol
Molecular FormulaC8H9ClO
InChI KeyDDUBOVLGCYUYFX-UHFFFAOYNA-N
SMILESCC(O)C1=CC=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥97.5%
It is an intermediate in organic syntheses and used as a pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an intermediate in organic syntheses and used as a pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents. Store under room tempera
RUO – Research Use Only

General References:

  1. John E. Arrowsmith.; Simon F. Campbell.; Peter E. Cross.; John K. Stubbs.; Roger A. Burges.; Donald G. Gardiner.; Kenneth J. Blackburn. Long-acting dihydropyridine calcium antagonists. 1. 2-Alkoxymethyl derivatives incorporating basic substituents. J. Med. Chem. 1986, 29 (9),1696-1702.
  2. Mary Beth Carter.;Birgit Schiott.; Alberto Gutierrez.; Stephen L. Buchwald. Enantioselective Hydrosilylation of Ketones with a Chiral Titanocene Catalyst.J. Am. Chem. Soc. 1994, 116 (26),11667-11670.