Thermo Scientific Chemicals

N-[4-(4-Fluorophenyl)-5-formyl-6-isopropyl-2-pyrimidinyl]-N-methylmethanesulfonamide, 99%, Thermo Scientific Chemicals

Catalog number: H61129.14
25 g, Each
Thermo Scientific Chemicals

N-[4-(4-Fluorophenyl)-5-formyl-6-isopropyl-2-pyrimidinyl]-N-methylmethanesulfonamide, 99%, Thermo Scientific Chemicals

Catalog number: H61129.14
25 g, Each
Quantity
Catalog number: H61129.14
also known as H61129-14
Price (USD)
Price: 107.00
Online price: 91.65
Your price:
Quantity
-

Chemical Identifiers

CAS
147118-37-4
IUPAC Name
N-[4-(4-fluorophenyl)-5-formyl-6-(propan-2-yl)pyrimidin-2-yl]-N-methylmethanesulfonamide
Molecular Formula
C16H18FN3O3S
InChI Key
WOCOTUDOVSLFOB-UHFFFAOYSA-N
SMILES
CC(C)C1=NC(=NC(C2=CC=C(F)C=C2)=C1C=O)N(C)S(C)(=O)=O
Assay (HPLC)
≥97.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text