Thermo Scientific Chemicals

5-Fluoronicotinic acid, 98%, Thermo Scientific Chemicals

Catalog number: H61162.22
100 g, Each
Thermo Scientific Chemicals

5-Fluoronicotinic acid, 98%, Thermo Scientific Chemicals

Catalog number: H61162.22
100 g, Each
Quantity
Catalog number: H61162.22
also known as H61162-22
Price (USD)

Chemical Identifiers

CAS
402-66-4
Assay (HPLC)
>97.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Fluoronicotinic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Soluble in methanol.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text