Methyl 2-fluoroacrylate, 95%, stab. with 1% BHT
Methyl 2-fluoroacrylate, 95%, stab. with 1% BHT
Methyl 2-fluoroacrylate, 95%, stab. with 1% BHT
Methyl 2-fluoroacrylate, 95%, stab. with 1% BHT
Thermo Scientific Chemicals

Methyl 2-fluoroacrylate, 95%, stab. with 1% BHT

CAS: 2343-89-7 | C4H5FO2 | 104.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H61343.14
also known as H61343-14
Price (EUR)
341,00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
341,00
Each
Add to cart
Chemical Identifiers
CAS2343-89-7
IUPAC Namemethyl 2-fluoroprop-2-enoate
Molecular FormulaC4H5FO2
InChI KeyZTZJVAOTIOAZGZ-UHFFFAOYSA-N
SMILESCOC(=O)C(F)=C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>94.0%
Methyl 2-fluoroacrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-fluoroacrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Slightly soluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed.
RUO – Research Use Only

General References:

  1. Hisanaka Ito; Akio Saito; Takeo Taguchi. Asymmetric Diels-Alder reactions of 2-fluoroacrylic acid derivatives. Part 1: The construction of fluorine substituted chiral tertiary carbon. Tetrahedron: Asymmetry. 1998, 9(11), 1979-1987.
  2. Vladimir S. Yarmolchuk; Pavel K. Mykhailiuk; Igor V. Komarov. Synthesis of β-fluoro-β-proline. Tetrahedron: Asymmetry. 2011, 52(12), 1300-1302.