Methyl 2-amino-3-methylbenzoate, 98%
Methyl 2-amino-3-methylbenzoate, 98%
Methyl 2-amino-3-methylbenzoate, 98%
Thermo Scientific Chemicals

Methyl 2-amino-3-methylbenzoate, 98%

CAS: 22223-49-0 | C9H11NO2 | 165.192 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number H61448.14
also known as H61448-14
Price (USD)
67.65
Special offer
Online exclusive
Ends: 30-Jun-2026
79.20
Save 11.55 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
67.65
Special offer
Online exclusive
Ends: 30-Jun-2026
79.20
Save 11.55 (15%)
Each
Chemical Identifiers
CAS22223-49-0
IUPAC Namemethyl 2-amino-3-methylbenzoate
Molecular FormulaC9H11NO2
InChI KeyVSFYTPXXMLJNAU-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=CC(C)=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow
FormLiquid
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only