5-Chloromethyl-1H-tetrazole, 95%
5-Chloromethyl-1H-tetrazole, 95%
5-Chloromethyl-1H-tetrazole, 95%
Thermo Scientific Chemicals

5-Chloromethyl-1H-tetrazole, 95%

CAS: 55408-11-2 | C2H3ClN4 | 118.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H61643.03
also known as H61643-03
Price (TWD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS55408-11-2
IUPAC Name5-(chloromethyl)-2H-1,2,3,4-tetrazole
Molecular FormulaC2H3ClN4
InChI KeyAGWSISOYPHROLN-UHFFFAOYSA-N
SMILESClCC1=NNN=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only