Thermo Scientific Chemicals

Nalpha,Nepsilon-Di-Boc-L-lysine N-succinimidyl ester, 97%, Thermo Scientific Chemicals

Catalog number: H62068.06
5 g, Each
Thermo Scientific Chemicals

Nalpha,Nepsilon-Di-Boc-L-lysine N-succinimidyl ester, 97%, Thermo Scientific Chemicals

Catalog number: H62068.06
5 g, Each
Quantity
Catalog number: H62068.06
also known as H62068-06
Price (USD)
Price: 140.00
Online price: 119.65
Your price:
Quantity
-

Chemical Identifiers

CAS
30189-36-7
IUPAC Name
2,5-dioxopyrrolidin-1-yl (2S)-2,6-bis({[(tert-butoxy)carbonyl]amino})hexanoate
Molecular Formula
C20H33N3O8
InChI Key
IQVLXQGNLCPZCL-UGPWUYPHNA-N
SMILES
CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(=O)ON1C(=O)CCC1=O
Assay (HPLC)
>96.0%

Description

Nalpha, Nepsilon-Di-Boc-L-lysine N-succinimidyl ester is used as a local anesthetic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nalpha, Nepsilon-Di-Boc-L-lysine N-succinimidyl ester is used as a local anesthetic.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text