1-Boc-N-Fmoc-D-tryptophan, 98%
1-Boc-N-Fmoc-D-tryptophan, 98%
1-Boc-N-Fmoc-D-tryptophan, 98%
Thermo Scientific Chemicals

1-Boc-N-Fmoc-D-tryptophan, 98%

CAS: 163619-04-3 | C31H30N2O6 | 526.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H62185.03
also known as H62185-03
Price (EUR)
87,30
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
87,30
Each
Chemical Identifiers
CAS163619-04-3
IUPAC Name(2R)-3-{1-[(tert-butoxy)carbonyl]-1H-indol-3-yl}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Molecular FormulaC31H30N2O6
InChI KeyADOHASQZJSJZBT-AREMUKBSSA-N
SMILESCC(C)(C)OC(=O)N1C=C(C[C@@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only