2-Bromo-4-fluorobenzylamine hydrochloride, 96%
2-Bromo-4-fluorobenzylamine hydrochloride, 96%
2-Bromo-4-fluorobenzylamine hydrochloride, 96%
Thermo Scientific Chemicals

2-Bromo-4-fluorobenzylamine hydrochloride, 96%

CAS: 289038-14-8 | C7H8BrClFN | 240.5 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H62200.06
also known as H62200-06
Price (USD)
688.65
Online Exclusive
765.00
Save 76.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
688.65
Online Exclusive
765.00
Save 76.35 (10%)
Each
Specifications
CAS289038-14-8
Chemical Name or Material2-Bromo-4-fluorobenzylamine hydrochloride
Formula Weight240.5
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
2-Bromo-4-fluorobenzylamine hydrochloride used as a intermediate in pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4-fluorobenzylamine hydrochloride used as a intermediate in pharmaceutical.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only
Lauren R. Donaldson; Stephen Wallace; David Haigh; E. Elizabeth Pattonc; and Alison N. Hulme. Rapid synthesis and zebrafish evaluation of a phenanthridine-based small molecule library. Org. Biomol. Chem. 2011, 92233-2239.