3-Cyano-4-fluorobenzeneboronic acid pinacol ester, 96%
3-Cyano-4-fluorobenzeneboronic acid pinacol ester, 96%
3-Cyano-4-fluorobenzeneboronic acid pinacol ester, 96%
Thermo Scientific Chemicals

3-Cyano-4-fluorobenzeneboronic acid pinacol ester, 96%

CAS: 775351-57-0 | C13H15BFNO2 | 247.076 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H62245.03
also known as H62245-03
Price (USD)
76.40
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
76.40
Each
Specifications
CAS775351-57-0
Chemical Name or Material3-Cyano-4-fluorobenzeneboronic acid pinacol ester
DOT InformationHazard Class: 6.1; Packaging Group: III
Formula Weight247.07
Health Hazard 1H301+H311+H331-H315-H319-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only