Thermo Scientific Chemicals

N-Fmoc-L-aspartic acid, 95%, Thermo Scientific Chemicals

Catalog number: H62274.14
25 g, Each
Thermo Scientific Chemicals

N-Fmoc-L-aspartic acid, 95%, Thermo Scientific Chemicals

Catalog number: H62274.14
25 g, Each
Quantity
Catalog number: H62274.14
also known as H62274-14
Price (USD)
Price: 182.00
Online price: 157.65
Your price:
Quantity
-

Chemical Identifiers

CAS
119062-05-4
IUPAC Name
2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanedioic acid
Molecular Formula
C19H17NO6
InChI Key
KSDTXRUIZMTBNV-UHFFFAOYNA-N
SMILES
OC(=O)CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Assay (HPLC)
>94.0%

Description

N-Fmoc-L-aspartic acid is used to biosynthesize other amino acids with in the human body.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-L-aspartic acid is used to biosynthesize other amino acids with in the human body.

Solubility
Soluble in methanol, and water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text