Specifications
CAS222530-33-8
Chemical Name or Materialcis-3-(Boc-amino)cyclohexanecarboxylic acid
Formula Weight243.29
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more