Thermo Scientific Chemicals

Methyl (S)-(+)-2-oxopyrrolidine-5-carboxylate, 97%, Thermo Scientific Chemicals

25g, Each
Thermo Scientific Chemicals

Methyl (S)-(+)-2-oxopyrrolidine-5-carboxylate, 97%, Thermo Scientific Chemicals

25g, Each
Quantity
Catalog number: H62543.14
also known as H62543-14
Price (USD)
Price: 266.00
Your Price:
Quantity
-

Chemical Identifiers

CAS
4931-66-2
Assay (unspecified)
>96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only