trans-2-(Benzyloxycarbonylamino)cyclohexanecarboxylic acid, 97%, Thermo Scientific Chemicals
trans-2-(Benzyloxycarbonylamino)cyclohexanecarboxylic acid, 97%, Thermo Scientific Chemicals
trans-2-(Benzyloxycarbonylamino)cyclohexanecarboxylic acid, 97%, Thermo Scientific Chemicals
trans-2-(Benzyloxycarbonylamino)cyclohexanecarboxylic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

trans-2-(Benzyloxycarbonylamino)cyclohexanecarboxylic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H62634.06
also known as H62634-06
Price (TWD)
-
Request A Quote
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS61935-48-6
IUPAC Name2-{[(benzyloxy)carbonyl]amino}cyclohexane-1-carboxylic acid
Molecular FormulaC15H19NO4
InChI KeyRPJMLWMATNCSIS-UHFFFAOYNA-N
SMILESOC(=O)C1CCCCC1NC(=O)OCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystalline powder
Assay (Titration)≥96.0%
Assay (HPLC)≥96.0%
Assay from Supplier's CofAcis- Isomer content: < 0.3%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only