4-Amino-3,5-difluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals
4-Amino-3,5-difluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals
4-Amino-3,5-difluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Amino-3,5-difluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H62654.03
also known as H62654-03
1 g
Catalog number H62654.03
also known as H62654-03
Price (EUR)
305,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
305,00
Each
Specifications
CAS939968-08-8
Chemical Name or Material4-Amino-3,5-difluorobenzeneboronic acid pinacol ester
ColorWhite
Formula Weight255.07
Health Hazard 1H315-H319-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only