N-Boc-o-tert-butil-L-treonina, 95 %, Thermo Scientific Chemicals
N-Boc-o-tert-butil-L-treonina, 95 %, Thermo Scientific Chemicals
N-Boc-o-tert-butil-L-treonina, 95 %, Thermo Scientific Chemicals
N-Boc-o-tert-butil-L-treonina, 95 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-o-tert-butil-L-treonina, 95 %, Thermo Scientific Chemicals

CAS: 13734-40-2 | C13H25NO5 | 275.35 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
1 g
5 g
Número de catálogo H62713.06
también denominado H62713-06
Precio (USD)
-
Cantidad:
5 g
Pedido a granel o personalizado
Identificadores químicos
CAS13734-40-2
IUPAC Name(2S,3R)-3-(tert-butoxy)-2-{[(tert-butoxy)carbonyl]amino}butanoic acid
Molecular FormulaC13H25NO5
InChI KeyLKRXXARJBFBMCE-BDAKNGLRSA-N
SMILESC[C@@H](OC(C)(C)C)[C@H](NC(=O)OC(C)(C)C)C(O)=O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Assay (unspecified)>94.0%
N-Boc-O-tert-butyl-L-threonine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Aplicaciones
La N-Boc-o-tert-butil-L-treonina se utiliza como intermediario farmacéutico.

Solubilidad
Soluble en DMF.

Notas
Almacenar en un lugar fresco y seco, en un recipiente bien sellado. Almacenar lejos de agentes oxidantes.
RUO – Research Use Only
  1. Antonio Procopio; Renato Dalpozzo; Antonio De Nino; Loredana Maiuolo; Monica Nardi; Manuela Oliverio; Beatrice Russo. A facile Er(OTf)3-catalyzed synthesis of 2,3-unsaturated O- and S-glycosides. Carbohydrate Research. 2007, 342 (14), 2125-2131.