Methyl 3-amino-5-fluorobenzoate, 98%
Methyl 3-amino-5-fluorobenzoate, 98%
Methyl 3-amino-5-fluorobenzoate, 98%
Thermo Scientific Chemicals

Methyl 3-amino-5-fluorobenzoate, 98%

CAS: 884497-46-5 | C8H8FNO2 | 169.155 g/mol
Have Questions?
Catalog NumberQuantity
H63038.03
also known as H63038-03
1 g
Catalog number H63038.03
also known as H63038-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS884497-46-5
IUPAC Namemethyl 3-amino-5-fluorobenzoate
Molecular FormulaC8H8FNO2
InChI KeyYKEJVXMOMKWEPV-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC(N)=CC(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
FormSolid
Assay (HPLC)>97.5%
Methyl 3-amino-5-fluorobenzoate is an amino acid widely used in pharmaceuticals and foods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-amino-5-fluorobenzoate is an amino acid widely used in pharmaceuticals and foods.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only